Merck
  • Cytoplasmic poly(ADP-ribose)polymerase from mouse plasmacytoma free messenger ribonucleoprotein particles: purification and characterization.
Cytoplasmic poly(ADP-ribose)polymerase from mouse plasmacytoma free messenger ribonucleoprotein particles: purification and characterization.
Biochemical and biophysical research communications (1993-09-15)
M Jesser, C Chypre, F Hog, P Mandel
ABSTRAKT
A cytoplasmic poly(ADP-ribose)polymerase (PARP) was purified from mouse plasmacytoma free messenger ribonucleoprotein particles using chromatography on 3-aminobenzamide affigel-10. The purified protein showed one band at 116 kDa on SDS-polyacrylamide gel electrophoresis and shared similar antigenic sites to the nuclear PARP. An apparent Km for NAD of 100.5 +/- 6.3 microM and a Vmax of 174 +/- 40 nmoles of ADP-ribose incorporated/min/mg protein were observed. RNA was detected in the enzyme preparation and the enzymatic activity was not DNA dependent.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3-Methoxybenzamide, 97%