Merck
  • The luteal phase after ovulation induction with human menopausal gonadotropin and one versus two doses of a gonadotropin-releasing hormone agonist.
The luteal phase after ovulation induction with human menopausal gonadotropin and one versus two doses of a gonadotropin-releasing hormone agonist.
Fertility and sterility (1993-06-01)
S L Corson, F R Batzer, B Gocial, G Maislin
ABSTRAKT
To use a GnRH agonist (GnRH-a) to induce ovulation after priming with exogenous hMG. Prospective, randomized double-blind protocol using one or two doses of intranasal nafarelin. Office-based ovulation induction program. PATIENTS, INTERVENTIONS: Infertile women not conceiving after use of clomiphene citrate for at least 6 months who were given hMG and nafarelin. No luteal support was given. Serum concentrations of FSH, LH, E2, and P acutely and at 6 days after GnRH-a administration. Duration of the luteal phase was assessed. Ovulation with elevation of both FSH and LH was achieved. The two-dose regimen was more effective than one dose for sustained LH release. Luteal phase P values and luteal phase duration were both less than usually seen with gonadotropin hCG therapy in the absence of luteal phase support. Ovulation induction with GnRH-a after hMG priming produces unacceptable luteal phase cycles in the absence of hormonal support.