Merck
In vitro effects of gamma-hexachlorocyclohexane on in vitro biosynthesis and metabolism of steroids in goldfish Carassius auratus.
Ecotoxicology and environmental safety (1996-07-01)
D E Kime, P B Singh
ABSTRAKT
Testicular and ovarian fragments of Carassius auratus, taken during the reproductively active prespawning phase (June) of its annual reproductive cycle, were incubated with different concentrations (0 mg/ml, 0 ppm; 0.001 mg/ml, 1 ppm; 0.01 mg/ml, 10 ppm; and 0.02 mg/ml, 20 ppm) of gamma-hexachlorocyclohexane (gamma-HCH) in the presence of either exogenous precursor [3H]-17-hydroxyprogesterone ([3H]17-P) or carp hypophyseal homogenate. The free (unconjugated) and conjugated metabolites (glucuronides and sulfates) of [3H]-17-P [androstenedione (AD), androstenetrione, 17-hydroxyprogesterone, testosterone (T), 11-deoxycortisol (S), 17, 20alpha-dihydroxy-4-pregnen-3-one (17,20alphaP), 17, 20beta-dihydroxy-4-pregnen-3-one (17,20betaP), 7alpha-pregnanetetrols (7alpha-P), and other polar metabolites] were separated by thin-layer chromatography and high-performance liquid chromatography. The endogenous production of unconjugated (free) steroids T, 17,20betaP, S, and 11-ketotestosterone (11-KT) in response to gamma-HCH were measured by radioimmunoassay. Among the in vitro metabolism of [3H]-17-P, in males, free steroids of AD, T, 17,20alphaP, S, and polar-free steroids were increased with the decreased yield of 11-KT. Percentage yield of testosterone glucuronide (TG) was increased with highly significant decreased yields of polar glucuronide steroids. The sulfate steroids of 17, 20alphaP, 17,20betaP, S, and 11-KT remain unchanged. In females, the decreased percentage of yield of AD and S and elevated T were noticed. The yield of TG was increased with decreased yield of 7alpha-P glucuronides. The percentage yield of AD sulfate and sulfate steroids of 17,20alphaP, 17,20betaP, and S were noted to be increased, but the yield for S sulfate was very high. Endogenous production of T was increased in both sexes in the presence of gamma-HCH, but 11-KT in male and S in female were depressed. 17, 20betaP was stimulated at some concentrations in both sexes but levels were very low. Results indicate that gamma-HCH in vitro perturbed the steroid biosynthesis during this phase thereby affecting reproductive physiology.