Merck
Simultaneous high-performance liquid chromatographic analysis of flunitrazepam and four metabolites in serum.
Journal of chromatography. B, Biomedical applications (1996-10-25)
F Berthault, P Kintz, P Mangin
ABSTRAKT
A high-performance liquid chromatographic method for the simultaneous determination of flunitrazepam and four metabolites, desmethylflunitrazepam (DMF), 7-aminodesmethylflunitrazepam (7-NH2DMF), 7-aminoflunitrazepam (7-NH2F) and 3-hydroxyflunitrazepam (3-OHF), in serum is described. The method involves a simple extraction from alkalinized plasma (pH 9.5) into diethyl ether-chloroform (80:20, v/v). Prazepam was used as an internal standard for the quantification of the five compounds. Separation was achieved with a 10 microns RSil CN column (300 x 3.9 mn I.D.). The detection wavelength was set at 242 nm. The limits of detection ranged from 2.5 to 5 micrograms/l with a limit of quantification of 10 micrograms/l for all analytes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Prazepam
Supelco
Prazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®