Merck
  • [3H]inositol polyphosphate metabolism in muscarinic cholinoceptor-stimulated airways smooth muscle: accumulation of [3H]inositol 4,5 bisphosphate via a lithium-sensitive inositol polyphosphate 1-phosphatase.
[3H]inositol polyphosphate metabolism in muscarinic cholinoceptor-stimulated airways smooth muscle: accumulation of [3H]inositol 4,5 bisphosphate via a lithium-sensitive inositol polyphosphate 1-phosphatase.
The Journal of pharmacology and experimental therapeutics (1997-02-01)
B J Lynch, M M Muqit, T R Walker, E R Chilvers
ABSTRAKT
Agonist-stimulated phosphoinositide hydrolysis is the principal mechanism underlying pharmacomechanical coupling in airways smooth muscle. In bovine tracheal smooth muscle, activation of muscarinic cholinoceptors results in sustained phospholipase C-mediated PtdIns(4,5)P2 hydrolysis but transient Ins(1,4,5)P3 accumulation, which implies agonist-stimulated metabolism of Ins(1,4,5)P3. To investigate the metabolic fate of Ins(1,4,5)P3 in bovine tracheal smooth muscle, we developed a [3H]inositol-labeling protocol wherein more than 98% of the [3H]inositol polyphosphates that accumulated over a 0 to 30-min incubation with 100 microM carbachol in the presence of 5 mM LiCl were derived from [3H]Ins(1,4,5)P3 and wherein the Ins(1,4,5)P3 3-kinase (EC 2.7.1.127) and 5-phosphatase (EC 3.1.3.56) pathways generated a set of mutually exclusive [3H]-inositol polyphosphate isomers. Under these conditions, the 5-phosphatase pathway was shown to be the dominant route for [3H]Ins(1,4,5)P3 metabolism at all time intervals measured, especially at early times (0-300 sec), where it accounted for more than 85% of [H]Ins(1,4,5)P3 metabolism. We also observed accumulation of a novel agonist and LiCl-sensitive [3H]InsP2 isomer identified as [3H]Ins(4,5)P2. The presence of a LiCI-sensitive inositol polyphosphate 1-phosphatase (EC 3.1.3.57) was demonstrated, and high LiCl concentrations (30 mM) caused a significant enhancement of [3H]Ins(1,4)P2 accumulation and a corresponding decline in [3H]Ins4P levels. Because nearly identical bell-shaped LiCl concentration-response curves were obtained for [H]Ins4P and [3H]Ins(4,5)P2 accumulation, and [3H]Ins(4,5)P2 was not generated under conditions expected to stimulate phospholipase D, these data suggest that the most likely precurser of [3H]Ins(4,5)P2 is [3H]Ins(1,4,5)P3. This is the first demonstration of Ins(4,5)P2 accumulation in a non-neuronal cell type, and the foregoing data suggest a novel route of formation via an Ins(1,4,5)P3 1-phosphatase, which would represent an additional pathway for [H]Ins(1,4,5)P3 removal.