Merck
Actions of diaspirin cross-linked hemoglobin on isolated rat and dog vessels.
The Journal of laboratory and clinical medicine (1997-03-01)
J L Hart, M A Ledvina, S M Muldoon
ABSTRAKT
The objectives of these studies were to investigate the responses of isolated blood vessels from rats and dogs to the administration of diaspirin cross-linked hemoglobin (DCLHb) and to determine the mechanisms of these responses. Isolated vascular rings (3 to 5 mm) were suspended at optimal passive tension in Krebs-filled (37 degrees C) tissue baths and bubbled with 95% O2-5% CO2, and isometric tension was recorded. With the vessels under basal conditions increasing concentrations of DCLHb (10(-8)-3 x 10(-6) mol/L) were added. DCLHb addition was repeated during a submaximal contraction with norepinephrine and again during acetylcholine relaxation. The effects of the nitric oxide synthase inhibitor L-nitro arginine (10(-5) mol/L) on the responses to DCLHb were also determined. Dog vessels developed very little tension (1% to 5% of norepinephrine maximum), whereas rat arteries contracted between 9% and 15% when exposed to DCLHb under basal conditions. However, both the dog and rat vessels developed significant tension to DCLHb when they were precontracted (5% to 54%) and also when they were relaxed with acetylcholine (21% to 93%). L-nitro arginine eliminated the contractile responses to DCLHb but did not cause contraction of any of the vessels under basal conditions. We conclude that in this model the mechanism of DCLHb-induced contractions of in vitro dog and rat vessels is dependent on interference with nitric oxide. This is similar to the mechanism of DCLHb action in isolated pig vessels reported previously. Differences in responses of dog, rat, and pig vessels under basal conditions in vitro are the result of active generation of nitric oxide by pig but not by dog or rat vessels.