Merck
Synthesis and antiparasitic activities of amidinic azolated derivatives.
Farmaco (Societa chimica italiana : 1989) (1997-04-01)
A Danan, D Charon, S Kirkiacharian, C Bories, P M Loiseau
ABSTRAKT
A set of heterocyclic N-acetamidinium hydrochlorides were prepared from the corresponding N-acetonitriles. The antiparasitic screening showed that, while all amidines are practically inactive, some nitriles present leishmanicide properties.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetamidine hydrochloride, 95%