Merck
Effect of IBMX and alkaline phosphatase inhibitors on Cl- secretion in G551D cystic fibrosis mutant mice.
The American journal of physiology (1998-03-05)
S N Smith, S J Delaney, J R Dorin, R Farley, D M Geddes, D J Porteous, B J Wainwright, E W Alton
ABSTRAKT
Some cystic fibrosis transmembrane conductance regulator (CFTR) mutations, such as G551D, result in a correctly localized Cl- channel at the cell apical membrane, albeit with markedly reduced function. Patch-clamp studies have indicated that both phosphatase inhibitors and 3-isobutyl-1-methylxanthine (IBMX) can induce Cl- secretion through the G551D mutant protein. We have now assessed whether these agents can induce Cl- secretion in cftrG551D mutant mice. No induction of Cl- secretion was seen with the alkaline phosphatase inhibitors bromotetramisole or levamisole in either the respiratory or intestinal tracts of wild-type or cftrG551D mice. In contrast, in G551D intestinal tissues, IBMX was able to produce a small CFTR-related secretory response [means +/- SE: jejunum, 1.8 +/- 0.9 microA/cm2, n = 7; cecum, 3.7 +/- 0.8 microA/cm2, n = 7; rectum (in vivo), 1.9 +/- 0.9 mV, n = 5]. This was approximately one order of magnitude less than the wild-type response to this agent and, in the cecum, was significantly greater than that seen in null mice (cftrUNC). In the trachea, IBMX produced a transient Cl- secretory response (37.3 +/- 14.7 microA/cm2, n = 6) of a magnitude similar to that seen in wild-type mice (33.7 +/- 4.7 microA/cm2, n = 9). This response was also present in null mice and therefore is likely to be independent of CFTR. No effect of IBMX on Cl- secretion was seen in the nasal epithelium of cftrG551D mice. We conclude that IBMX is able to induce detectable levels of CFTR-related Cl- secretion in the intestinal tract but not the respiratory tract through the G551D mutant protein.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(−)-p-Bromolevamisole oxalate, 99%