Merck
Allergic contact dermatitis from naftifine in a child without cross-reaction to terbinafine.
Journal of the European Academy of Dermatology and Venereology : JEADV (1998-09-10)
J J Goday, M González-Güemes, I Yanguas, R Ilardia, R Soloeta
ABSTRAKT
Allergic contact dermatitis from naftifine has been scarcely described in the English literature, all of them in adults. We report a case of a 12-year-old girl who developed an acute eczema on her neck after application of a naftifine cream. This fact was confirmed by a patch-test study. We did not find a cross-reaction to terbinafine, a structurally linked allylamine.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Naftifine hydrochloride