Merck
[Antigen-induced airway hyperresponsiveness in infantile guinea pigs].
Arerugi = [Allergy] (1998-10-31)
H Taniguchi, G Tamura, H Iijima, Y Liu, K Shirato
ABSTRAKT
To investigate the development of airway hyperresponsiveness in infantile guinea pigs, animals (10 days old) were immunized twice and challenged by inhalation of 1% ovalbumin for 10 min with 7 days intervals. Similar to adult guinea pigs, infantile ones developed an increased airway responsiveness to acetylcholine 24 hr after antigen challenge. There was a marked increase in the number of total leukocytes, eosinophils and lymphocytes in bronchoalveolar lavage fluid (BALF). Suplatast tosilate (suplatast) and pemirolast potassium (pemirolast) given orally throughout the experiments suppressed the development of airway hyperresponsiveness in infantile animals. They showed similar potency in the suppression of eosinophil accumulation in BALF and lung tissue, while suplatast inhibited lymphocyte accumulation stronger than pemirolast. Collectively, the present model of airway hyperresponsiveness in infantile guinea pigs may be useful in predicting the efficacy of antiallergic agents in the treatment of asthmatic children.