Merck
Serum amyloid A stimulates lipoprotein-associated phospholipase A2 expression in vitro and in vivo.
Atherosclerosis (2013-04-30)
Bo Li, Zhe Dong, Hui Liu, Yan-Fei Xia, Xiao-Man Liu, Bei-Bei Luo, Wen-Ke Wang, Bin Li, Fei Gao, Cheng Zhang, Ming-Xiang Zhang, Yun Zhang, Feng-Shuang An
ABSTRAKT
Although lipoprotein-associated phospholipase A2 (Lp-PLA2) has been regarded as a biomarker and a causative agent for acute coronary events recently, the mechanism of the regulation of Lp-PLA2 has not been fully elucidated yet. This study was aimed to investigate the influence of serum amyloid A (SAA) on the expression of Lp-PLA2 in THP-1 cells and ApoE-deficient (ApoE(-/-)) mice. THP-1 cells were stimulated by SAA and the mRNA and protein expression of Lp-PLA2 was detected. ApoE(-/-) mice were intravenously injected with murine SAA1 lentivirus. Formyl peptide receptor like-1 (FPRL1) agonist (WKYMVm) and inhibitor (WRW(4)), mitogen-activated protein kinases (MAPKs) inhibitors, and peroxisome proliferator-activated receptor-γ (PPAR-γ) agonist and inhibitor were used to investigate the mechanism of regulation of Lp-PLA2. Recombinant SAA up-regulated Lp-PLA2 expression in a dose and time-dependent manner in THP-1 cells. Immunohistochemical analysis of aortic root of ApoE(-/-) mice also demonstrated that the expression of Lp-PLA2 was up-regulated significantly with SAA treatment. WRW(4) decreased SAA-induced Lp-PLA2 production; while WKYMVm could induce Lp-PLA2 expression. ERK1/2, JNK1/2, and p38 inhibition reduced SAA-induced Lp-PLA2 production. Furthermore, the results suggested the activation of PPAR-γ played a crucial role in this process. These results demonstrate that SAA up-regulates Lp-PLA2 production significantly via a FPRL1/MAPKs./PPAR-γ signaling pathway.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
WKYMVM trifluoroacetate salt, ≥98% (HPLC)