Merck
Inhibition of proliferation of contaminating fibroblasts by D-valine in cultures of smooth muscle cells from human myometrium.
Cell biology international (2000-05-29)
J Hongpaisan
ABSTRAKT
Replacement of L-valine with D-valine in a standard culture medium can selectively inhibit fibroblast proliferation. The aim of the present study was to investigate whether human myometrial cells cultured with D-valine instead of L-valine can survive and express their characteristics. Cultured cells (95-98%) maintain expression of the intermediate filament desmin, which is the specific marker for mature muscle cells. By transmission electron microscopy, the cells showed the general morphology of smooth muscle cells in culture. Oxytocin in serum-free culture medium at 37 degrees C (5 min) caused a concentration-dependent increase in cellular Na and total Ca, and a decrease in K content as determined by X-ray microanalysis. The percentage of cells cultured with D-valine responding to oxytocin stimulation was larger than that of cells cultured with L-valine, suggesting less contamination of smooth muscle cells by fibroblasts in the presence of D-valine. As shown by measurements with fura-2, D-valine-cultured cells retained the characteristic increase in intracellular free Ca2+ ions after oxytocin stimulation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Valine
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)