Merck
Detection of rare malignant cells and their apoptotic fragments in cerebrospinal fluid.
Lancet (London, England) (2000-11-10)
B R Kranz
ABSTRAKT
Routine cerebrospinal fluid cytology often fails to detect small numbers of malignant cells exfoliated from primary lymphomas of the central nervous system and metastatic carcinomas. Immunocytochemistry on poly-L-lysine-coated slides was optimised to permit unequivocal identification of a single carcinoma cell in 1 mL of cerebrospinal fluid, as well as carcinoma cell-derived apoptotic bodies that themselves contribute to enhanced diagnostic sensitivity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)