- Oxygen administration in infants.
Oxygen administration in infants.
Archives of disease in childhood. Fetal and neonatal edition (2003-02-25)
B Frey, F Shann
PMID12598492
ABSTRAKT
The main methods of oxygen administration to infants are reviewed. Some methods are more economical and therefore more useful in developing countries. All the methods have potential complications and therefore need to be carefully supervised.