Merck
The reduction of oxo-acids by human tissue extracts.
Clinical biochemistry (1976-04-01)
F Naghizadeh, D Barlow, J King
ABSTRAKT
The reduction of pyruvate, 2-oxobutyrate, hydroxy-pyruvate and glyoxylate under optimal conditions at 37 degrees by extracts of human heart, kidney, pancreas, skeletal muscle and liver has been examined. The ratios of reduction rates, particularly of glyoxylate and 2-oxobutyrate reveal a capacity to differentiate the isoenzyme composition of the tissue extracts.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glyoxylic acid solution, 50 wt. % in H2O