Merck
The status of plasma homocysteine and related B-vitamins in healthy young vegetarians and nonvegetarians.
European journal of nutrition (2003-03-15)
Yi-Chia Huang, Sue-Joan Chang, Yu-Ting Chiu, Han-Hsin Chang, Chien-Hsiang Cheng
ABSTRAKT
Exclusion of animal products and having only plant protein in vegetarian diets may affect the status of certain B-vitamins, and further cause the elevation of plasma homocysteine concentration. The purpose of this study was to assess the status of homocysteine and related B-vitamins in vegetarians and nonvegetarians. The effects of biochemical parameters of B-vitamins and dietary protein on plasma homocysteine were also examined. The study was performed at the Chung Shan Medical University, Taichung, in the central part of Taiwan. Thirty-seven vegetarians (28.9 +/- 5.5 y) and 32 nonvegetarians (22.9 +/- 1.6 y) were recruited. Nutrient intake was recorded using 3-day dietary records. Fasting venous blood samples were obtained. Plasma homocysteine, folate and vitamin B-12 were measured. Vitamin B-6 status was assessed by direct measures [plasma pyridoxal 5'-phosphate (PLP) and urinary 4-pyridoxic acid (4-PA)] and indirect measures [erythrocyte alanine (EALT-AC) and aspartate (EAST-AC) aminotransaminase activity coefficient]. There was no significant difference in vitamin B-6 intake between the two groups, although the vegetarian group had a significantly lower vitamin B-12 intake than the nonvegetarian group. Vegetarian subjects had significantly lower mean plasma PLP and vitamin B-12 concentrations than did nonvegetarian subjects (p < 0.05); however, a significantly higher mean plasma folate concentration was found in the vegetarian group. Vegetarian subjects had a significantly higher mean plasma homocysteine concentration than nonvegetarian subjects (13.2 +/- 7.9 vs. 9.8 +/- 2.2 micromol/L). Negative correlations were seen between plasma homocysteine and vitamin B-12 concentrations in the vegetarian (p = 0.004), nonvegetarian (p = 0.026), and pooled (p < 0.001) groups. From best subsets regression analyses, the plasma homocysteine concentration could be significantly predicted by total protein intake (p = 0.027) and plasma vitamin B-12 concentration (p = 0.005) in the pooled group. When the intake of protein is not considered, vitamin B-12 concentration is still a strong predictor of plasma homocysteine concentration (p = 0.012). Vitamin B-12 intake and mean plasma vitamin B-12 concentration were lower for vegetarian subjects than for nonvegetarian subjects, leading to an increase in plasma homocysteine concentration. Vitamin B-6 and folate had little effect on plasma homocysteine concentration when individuals had adequate vitamin B-6 and folate status.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Pyridoxic acid, ≥98%