Merck
Neurochemical effects of vagus nerve stimulation in humans.
Brain research (1992-06-26)
E J Hammond, B M Uthman, B J Wilder, E Ben-Menachem, A Hamberger, T Hedner, R Ekman
ABSTRAKT
An implanted stimulating device chronically stimulated the left cervical vagus nerve in epileptic patients. Cerebrospinal fluid concentrations of free and total gamma-aminobutyric acid, homovanillic acid, 5-hydroxyindoleacetic acid, aspartate, glutamate, asparagine, serine, glutamine, glycine, phosphoethanolamine, taurine, alanine, tyrosine, ethanolamine, valine, phenylalanine, isoleucine, vasoactive intestinal peptide, beta-endorphin, and somatostatin were measured before and after 2 months of chronic stimulation in six patients. Significant increases were seen in homovanillic acid and 5-hydroxyindoleacetic acid in three patients, and significant decreases in aspartate were seen in five patients. These changes were associated with a decrease in seizure frequency.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
Supelco
Ethanolamine, analytical standard