Merck
Absorption, metabolism, degradation and urinary excretion of rosmarinic acid after intake of Perilla frutescens extract in humans.
European journal of nutrition (2004-08-17)
Seigo Baba, Naomi Osakabe, Midori Natsume, Akiko Yasuda, Yuko Muto, Kyoko Hiyoshi, Hirohisa Takano, Toshikazu Yoshikawa, Junji Terao
ABSTRAKT
Rosmarinic acid (RA) is a natural polyphenolic substance contained in many Lamiaceae herbs such as Perilla frutescens. Previous studies have shown RA has antioxidative and anti-inflammatory activity. However, little is known on the absorption, metabolism, degradation and excretion of RA. The aim of this study in healthy humans was to determine the absorption, metabolism, and urinary excretion of RA after a single intake of perilla extract (PE). Six healthy men (mean age 37.2 +/- 6.2 y and mean body mass index 22.0 +/- 1.9 kg/m(2)) were enrolled in the study that was a crossover design involving single intakes of PE containing 200 mg RA and placebo with a 10 day interval between treatments. Blood samples were collected before intake and at designated time intervals, while urine samples were collected over the periods 0-6 h, 6-24 h and 24-48 h after intake. RA and its related metabolites in plasma and urine were measured by LC-MS. RA, methylated RA (methyl-RA), caffeic acid (CAA), ferulic acid (FA) and a trace of m-coumaric acid (COA) were detected in the urine after intake of PE. In plasma, RA, methyl-RA and FA were detected, with maximum levels obtained 0.5, 2 and 0.5 h after intake of PE, respectively. The majority of these components in both plasma and urine were present as conjugated forms (glucuronide and/or sulfated). The proportion of RA and its related metabolites excreted in the urine was 6.3 +/- 2.2% of the total dose, with approximately 75% of these components being excreted within 6 h after intake of PE. RA contained in PE was absorbed, conjugated and methylated following intake, with a small proportion of RA being degraded into various components, such as conjugated forms of CAA, FA and COA. These metabolites were then rapidly excreted in the urine.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
trans-Ferulic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
trans-Ferulic acid, 99%