Merck
  • Development and validation of a liquid chromatographic/electrospray ionization mass spectrometric method for the determination of benazepril, benazeprilat and hydrochlorothiazide in human plasma.
Development and validation of a liquid chromatographic/electrospray ionization mass spectrometric method for the determination of benazepril, benazeprilat and hydrochlorothiazide in human plasma.
Journal of mass spectrometry : JMS (2006-03-17)
Ariadni Vonaparti, Michael Kazanis, Irene Panderi
ABSTRAKT
A new method was developed and fully validated for the quantitation of benazepril, benazeprilat and hydrochlorothiazide in human plasma. Sample pretreatment was achieved by solid-phase extraction (SPE) using Oasis HLB cartridges. The extracts were analysed by high-performance liquid chromatography (HPLC) coupled to a single-quadrupole mass spectrometer (MS) with an electrospray ionization interface. The MS system was operated in selected ion monitoring (SIM) modes. HPLC was performed isocratically on a reversed-phase porous graphitized carbon (PGC) analytical column (2.1 x 125.0 mm i.d., particle size 5 microm). The mobile phase consisted of 55% acetonitrile in water containing 0.3% v/v formic acid and pumped at a flow rate of 0.15 ml min(-1). Chlorthalidone was used as the internal standard (IS) for quantitation. The assay was linear over a concentration range of 5.0-500 ng ml(-1) for all the compounds analysed, with a limit of quantitation of 5 ng ml(-1) for all the compounds. Quality control (QC) samples (5, 10, 100 and 500 ng ml(-1)) in five replicates from three different runs of analyses demonstrated intra-assay precision (coefficient of variation (CV) < or =14.6%), inter-assay precision (CV < or = 5.6%) and overall accuracy (relative error less than -8.0%). The method can be used to quantify benazepril, benazeprilat and hydrochlorothiazide in human plasma, covering a variety of pharmacokinetic or bioequivalence studies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrochlorothiazide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrochlorothiazide, meets USP testing specifications
Sigma-Aldrich
Hydrochlorothiazide, crystalline
Supelco
Hydrochlorothiazide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®