Merck
Ursolic acid regulates high glucose-induced apoptosis.
Free radical research (2007-05-23)
Chang Joo Oh, In Sup Kil, Chan Ik Park, Chae Ha Yang, Jeen-Woo Park
ABSTRAKT
A high concentration of glucose has been implicated as a causal factor in initiation and progression of diabetic complications and there is evidence to suggest that hyperglycemia increases the production of free radicals and oxidative stress. Therefore, compounds that scavenge reactive oxygen species (ROS) may confer regulatory effects on high glucose-induced apoptosis. Ursolic acid (UA), a pentacyclic triterpene, is reported to have an antioxidant activity. We investigated the effect of UA on high glucose-induced apoptosis in U937 cells. Upon exposure to 35 mM glucose for two days, there was a distinct difference between untreated cells and cells pre-treated with 50 nM UA for 2 h in regard to cellular redox status and oxidative DNA damage to cells. UA pre-treated cells showed significant suppression of apoptotic features such as DNA fragmentation, damage to mitochondrial function and modulation of apoptotic marker proteins upon exposure to high glucose. This study indicates that UA may play an important role in regulating the apoptosis induced by high glucose presumably through scavenging of ROS.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ursolic acid, ≥90%
Supelco
Ursolic acid, analytical standard
Ursolic acid, primary reference standard