Merck
Phytanic acid and very long chain fatty acids in genetic peroxisomal disorders.
Journal of clinical chemistry and clinical biochemistry. Zeitschrift fur klinische Chemie und klinische Biochemie (1989-05-01)
B Molzer, M Kainz-Korschinsky, R Sundt-Heller, H Bernheimer
ABSTRAKT
1. Phytanic acid, phytanyl-triacylglycerols, and very long chain fatty acids were analysed by gas chromatography or thin-layer chromatography in blood and tissues of patients with different genetic peroxisomal disorders (Refsum's disease, X-linked adrenoleukodystrophy, neonatal adrenoleukodystrophy, Zellweger syndrome). 2. We evaluated these analyses in the detection of patients with Refsum's disease, X-linked adrenoleukodystrophy, neonatal adrenoleukodystrophy, and Zellweger syndrome, and of carriers of X-linked adrenoleukodystrophy. In particular, the analysis of phytanyl-triacylglycerols by thin-layer chromatography proved to be a rapid and reliable method for the detection of patients and the monitoring of their dietary treatment in Refsum's disease. In X-linked adrenoleukodystrophy, carrier detection may depend on very long chain fatty acid analysis in more than one material (e.g. plasma and fibroblasts). 3. Analysis of phytanic acid showed that in patients with multiple impairments of peroxisomal functions (Zellweger syndrome, neonatal adrenoleukodystrophy) phytanic acid levels may be increased not only in serum, but also in the tissue (e.g. brain, adrenals, kidney). 4. Analysis of very long chain fatty acids in cholesterol esters from the brain, adrenals, kidney, and liver of patients with peroxisomal disorders revealed four different types of very long chain fatty acid patterns according to the behaviour of C 26:0 and of other saturated and monounsaturated very long chain fatty acids.(ABSTRACT TRUNCATED AT 250 WORDS)
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phytanic acid, ≥96%, mixture of isomers
Sigma-Aldrich
Octacosanoic acid, synthetic, ≥98%