Merck
Birthweights in the Faroe Islands: possible role of isovaleric acid.
Journal of internal medicine (1989-02-01)
R G Ackman
ABSTRAKT
A high intake of n-3 polyunsaturated fatty acids has been suggested as a factor in prolonged gestation in the population of the Faroe Islands. It is now suggested that isovaleric acid from pilot whales, a species frequently consumed in the Faroe Islands, may be the unusual dietary factor. Fatty acid data for eicosapentaenoic acid (EPA) and docosahexanoic acid (DHA) in blood lipids of Faroese and Norwegians is reviewed in terms of the type of fish eaten, apparently mostly lean white fish with DHA much greater than EPA. The popular lean fish, thus, probably provides too little EPA to produce a marked effect on human biochemistry.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Isovaleric acid, natural, ≥98%, FG
Sigma-Aldrich
Isovaleric acid, ≥99%, FCC, FG
Supelco
Isovaleric acid, analytical standard
Sigma-Aldrich
Isovaleric acid, 99%