Merck
IgA-mediated clearance and tissue deposition of dinitrophenylated human serum albumin at various DNP:HSA ratios.
Molecular immunology (1988-08-01)
J M Epps, J O Phillips, J Mestecky
ABSTRAKT
Dinitrophenylated human serum albumin (DNP-HSA) with various DNP:HSA ratios was prepared by direct conjugation with dinitrobenzene sulfate or with a caproic acid spacer group (DNP-cap-HSA) using DNP-epsilon-amino-caproic acid N-hydroxy-succinimide ester. The substitution ratio and chemical linkage (presence of a spacer group) were shown to affect the degree of murine anti-DNP antibody binding to antigen, and hence the tissue deposition and efficiency of hepatobiliary transport. DNP-HSA (4.5:1), which poorly binds to IgA anti-DNP antibody, is inefficiently transported into mouse bile. In contrast, 9:1 DNP-cap-HSA readily forms complexes and is more effectively cleared by the hepatobiliary route. The IgA-mediated increase in liver deposition of DNP-cap-HSA (9:1) was found to be associated with an increase in antigen uptake by hepatocytes. In contrast, large complexes formed between DNP-HSA (49:1) and IgA anti-DNP antibody are taken up by nonparenchymal cells of the liver and thus are inefficiently transported into bile. These results suggest that the IgA-mediated uptake by phagocytic cells or hepatocytes of haptenated protein strongly depends on the degree of haptenation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hexanoic acid, ≥99%
Sigma-Aldrich
Hexanoic acid, purum, ≥98.0% (GC)
Supelco
Hexanoic acid, analytical standard
Sigma-Aldrich
Hexanoic acid, ≥98%, FCC, FG