Merck
  • The neurotoxin MPTP does not reproduce in the rhesus monkey the interregional pattern of striatal dopamine loss typical of human idiopathic Parkinson's disease.
The neurotoxin MPTP does not reproduce in the rhesus monkey the interregional pattern of striatal dopamine loss typical of human idiopathic Parkinson's disease.
Neuroscience letters (1988-10-05)
C Pifl, G Schingnitz, O Hornykiewicz
ABSTRAKT
Using high-pressure liquid chromatography with electrochemical detection, we measured dopamine (DA) and homovanillic acid (HVA) in caudate nucleus, putamen and substantia nigra in 4 untreated rhesus monkeys and 4 monkeys with permanent parkinsonism produced by repeated injections of 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP; total dose: 2.1-6.45 mg/kg, i.m.). MPTP consistently produced a severe striatal and nigral loss of DA and HVA and an increase in the ratio 'HVA/DA'. In this respect, MPTP mimicked the changes found in human Parkinson's disease (PD). However, MPTP lowered the DA in caudate (-99.6%) to the same degree as in putamen (-99.5%). This is in contrast to idiopathic PD where the caudate is significantly less affected by DA loss (-84%) than the putamen (-98%). Thus, in our rhesus monkeys MPTP failed to reproduce the interregional caudate-putamen gradient characteristic of idiopathic PD. The DA pattern produced by MPTP was similar to the DA loss in caudate (-98%) and putamen (-99%) observed in patients with postencephalitic parkinsonism.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent