Merck
  • Pyrimidine nucleotidase deficiency with active dephosphorylation of dTMP: evidence for existence of thymidine nucleotidase in human erythrocytes.
Pyrimidine nucleotidase deficiency with active dephosphorylation of dTMP: evidence for existence of thymidine nucleotidase in human erythrocytes.
Blood (1983-11-01)
D E Paglia, W N Valentine, A S Keitt, R A Brockway, M Nakatani
ABSTRAKT
Erythrocytes from a patient with classical pyrimidine nucleotidase (PyN) deficiency had less than 10% residual PyN activity with uridine 5'-monophosphate (UMP) or cytidine 5'-monophosphate (CMP) as substrate, but exhibited brisk nucleotidase activity with thymidine 5'-monophosphate (dTMP). This strongly suggests the existence of separate enzymes or isozymes of PyN in normal human erythrocytes--an hypothesis that should be tested by similar studies in other cases of severe PyN deficiency, whether induced by genetic defects or lead toxicity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cytidine 5′-monophosphate, Sigma Grade, ≥99% (HPLC), synthetic, powder