- High-performance liquid chromatography of alpha-keto acids in human saliva.
High-performance liquid chromatography of alpha-keto acids in human saliva.
Archives of oral biology (1983-01-01)
H Tsuchiya, I Hashizume, T Tokunaga, M Tatsumi, N Takagi, T Hayashi
PMID6581765
ABSTRAKT
alpha-Keto acids in human mixed saliva collected without stimulation were analysed by reversed-phase ion-pair high-performance liquid chromatography (HPLC). Several alpha-keto acids were found in saliva and their concentrations were: alpha-ketoglutaric acid (KGA), 221 +/- 142; pyruvic acid (PA), 7490 +/- 5600; alpha-ketoisovaleric acid (KIVA), 61 +/- 23; alpha-ketoisocaproic acid (KICA), 137 +/- 79; alpha-keto-beta-methylvaleric acid (KMVA), 41 +/- 19 nmol/dl (mean +/- SD, n = 40). Their levels proved to be lower than those in plasma, except that of PA. Their concentrations in saliva showed individual variation compared with those in blood.