Merck
Alterations of lymphocyte subsets in children of diabetic mothers.
Diabetologia (1994-11-01)
U Roll, J Scheeser, E Standl, A G Ziegler
ABSTRAKT
To investigate the impact of diabetic mothers on the maturation of the immune system in their offspring, immunophenotypic markers of major lymphocyte subpopulations were evaluated by two-colour flow cytometric analysis in 160 healthy children of diabetic mothers (100 with insulin-dependent diabetes mellitus (IDDM): 48 with gestational diabetes), including 22 neonates, 45 infants aged 8-12 months, 46 children aged 1-2 years, 29 children aged 3-6 years and 18 children aged 7-17 years. Results were compared with 21 neonates of healthy mothers from our hospital and with 110 paediatric subjects of a reference population. In neonates of diabetic mothers, percentages of total lymphocytes (p = 0.044), T and B lymphocytes (p = 0.004, respectively) were significantly decreased compared to our neonates of healthy mothers. By subdividing the group of neonates in offspring of mothers with IDDM (n = 15) or gestational diabetes (n = 7), differences compared to normal neonates were mainly observed in neonates of mothers with IDDM (T lymphocytes: p = 0.006; B lymphocytes: p = 0.008). In cord blood, 45.5% of neonates had antibodies to islet cells, insulin or glutamic acid decarboxylase, most likely transmitted through the placenta of the diabetic mother. No association was found between alterations of lymphocyte subsets and antibody-positivity in cord blood, nor was there any correlation of lymphocyte counts and mean HbA1 during pregnancy, maternal age at delivery, diabetes duration, or neonatal birth weight, respectively. Comparisons among age groups from newborn infants through adolescents revealed higher percentages of total lymphocytes and lower percentages of activated T cells in children of diabetic mothers compared to children of the reference population between the age of 1 to 6 years (67-73% of the cases above and 62-77% below the interquartiles of the reference range, respectively). No significant differences in lymphocyte subpopulations between children of mothers with IDDM diabetes and gestational diabetes have been detected. In addition, there were no abnormalities of lymphocyte subsets in children who are at high risk for the development of IDDM. In summary, we suggest that the observed changes in children of diabetic mothers may reflect a cellular immune reaction to the particular maternal environment, characterized by both an abnormal metabolic state and persisting autoimmunity in the affected mother.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)