Merck
  • Effects of lithium salts on the stereochemistry of ketone enolization by lithium 2, 2, 6, 6-tetramethylpiperidide (LiTMP). A convenient method for highly E-selective enolate formation.
Effects of lithium salts on the stereochemistry of ketone enolization by lithium 2, 2, 6, 6-tetramethylpiperidide (LiTMP). A convenient method for highly E-selective enolate formation.
Journal of the American Chemical Society (1991)
Hall PL, et al.