Merck
T lymphocytes promote the antiviral and inflammatory responses of airway epithelial cells.
PloS one (2011-10-25)
Lan Jornot, Samuel Cordey, Assunta Caruso, Christine Gerber, Marija Vukicevic, Caroline Tapparel, Laurent Kaiser, Danielle Burger, Eddy Roosnek, Jean Silvain Lacroix, Thierry Rochat
ABSTRAKT
T cells modulate the antiviral and inflammatory responses of airway epithelial cells to human rhinoviruses (HRV). Differentiated primary human nasal epithelial cells (HNEC) grown on collagen-coated filters were exposed apically to HRV14 for 6 h, washed thoroughly and co-cultured with anti-CD3/CD28 activated T cells added in the basolateral compartment for 40 h. HRV14 did not induce IFNγ, NOS2, CXCL8 and IL-6 in HNEC, but enhanced expression of the T cell attractant CXCL10. On the other hand, HNEC co-cultured with activated T cells produced CXCL10 at a level several orders of magnitude higher than that induced by HRV14. Albeit to a much lower degree, activated T cells also induced CXCL8, IL-6 and NOS2. Anti-IFNγ antibodies and TNF soluble receptor completely blocked CXCL10 upregulation. Furthermore, a significant correlation was observed between epithelial CXCL10 mRNA expression and the amounts of IFNγ and TNF secreted by T cells. Likewise, increasing numbers of T cells to a constant number of HNEC in co-cultures resulted in increasing epithelial CXCL10 production, attaining a plateau at high IFNγ and TNF levels. Hence, HNEC activation by T cells is induced mainly by IFNγ and/or TNF. Activated T cells also markedly inhibited viral replication in HNEC, partially through activation of the nitric oxide pathway. Cross-talk between T cells and HNEC results in activation of the latter and increases their contribution to airway inflammation and virus clearance.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture