Merck
Human CARD12 is a novel CED4/Apaf-1 family member that induces apoptosis.
Biochemical and biophysical research communications (2001-05-26)
B J Geddes, L Wang, W J Huang, M Lavellee, G A Manji, M Brown, M Jurman, J Cao, J Morgenstern, S Merriam, M A Glucksmann, P S DiStefano, J Bertin
ABSTRAKT
The CED4/Apaf-1 family of proteins functions as critical regulators of apoptosis and NF-kappaB signaling pathways. A novel human member of this family, called CARD12, was identified that induces apoptosis when expressed in cells. CARD12 is most similar in structure to the CED4/Apaf-1 family member CARD4, and is comprised of an N-terminal caspase recruitment domain (CARD), a central nucleotide-binding site (NBS), and a C-terminal domain of leucine-rich repeats (LRR). The CARD domain of CARD12 interacts selectively with the CARD domain of ASC, a recently identified proapoptotic protein. In addition, CARD12 coprecipitates caspase-1, a caspase that participates in both apoptotic signaling and cytokine processing. CARD12 may assemble with proapoptotic CARD proteins to coordinate the activation of downstream apoptotic and inflammatory signaling pathways.