Merck
The physiological deadlock between AMPK and gluconeogenesis: SOGA, a novel protein, may provide the key.
The American journal of pathology (2010-09-04)
Josephine M Forbes
ABSTRAKT
This Commentary discusses how suppressor of glucose by autophagy (SOGA) contributes to adiponectin-mediated insulin-dependent inhibition of autophagy during the activation of adenosine monophosphate kinase (AMPK).