Merck
Diethyl oxalate as new reagent for spectrophotometric determination of ketosteroids.
Analytical chemistry (1970-05-01)
S Görög
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Diethyl oxalate, ≥99%