Merck
N-Demethylation of dextromethorphan.
Journal of pharmaceutical sciences (1980-12-01)
N P Peet
ABSTRAKT
In addition to providing an efficient synthesis of 3-methoxymorphinan hydrochloride, the use of 2,2,2-trichloroethyl chloroformate in the N-demethylation of dextromethorphan led to the isolation of two novel zinc salts of 3-methoxymorphinan.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2,2,2-Trichloroethyl chloroformate, purum, ≥98.0% (NT)
Sigma-Aldrich
2,2,2-Trichloroethyl chloroformate, 98%