Merck
Water-participation in the crystalline-state photodimerization of cinnamic acid derivatives. A new type of organic photoreaction.
Bulletin of the Chemical Society of Japan (1976)
Nakanishi F, et al.