Merck
Synthesis, size control, and passivation of CdS nanoparticles in water/AOT/ <I>n</I>-heptane microemulsions.
Materials Science &amp; Engineering. C, Materials For Biological Applications (2003)
Caponetti E, et al.