Merck
POLY (VINYLBENZYLPHOSPHONIC ACID)-BASED PROTON CONDUCTORS AS POLYMER ELECTROLYTE MEMBRANES FOR FUEL CELLS APPLICATIONS.
Polymer Preprints (American Chemical Society, Division of Polymer Chemistry) (2008)