Merck
A temperature-sensitive drug release system based on phase-change materials.
Angewandte Chemie (International ed. in English) (2010-09-15)
Sung-Wook Choi, Yu Zhang, Younan Xia
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Tetradecanol, 97%