Merck
Transient alterations of the blood-brain barrier tight junction and receptor potential channel gene expression by chlorpyrifos.
Journal of applied toxicology : JAT (2012-05-23)
Wen Li, Marion Ehrich
ABSTRAKT
The blood-brain barrier (BBB) is formed by specialized endothelial cells lining capillaries in the central nervous system (CNS). We previously demonstrated that exposure to very low concentrations of the organophosphorus insecticide chlorpyrifos (CPF) decreased electrical resistance across the BBB in vitro, indicating a loss of BBB integrity. The present study examined the transient effects of CPF on expression of genes contributing to tight junctions of the BBB. Rat brain endothelial cells (RBE4) were co-cultured with rat astrocytes on membrane inserts to form an in vitro BBB. The RBE4 cells in the BBB were then exposed to CPF for 2, 4 and 12 h. Total RNA was extracted from RBE4 cells and quantitative real-time PCR (qRT-PCR) was used to quantify levels of gene expression of tight junction proteins claudin5, scaffold proteins zona occludens (ZO1) and transient receptor potential (canonical) channels (TRPC4). Gene expression decreased 2 h after exposure to CPF, especially TRPC4, but the effects were reversed 12 h later. CPF exposure for only 15 min caused less effect than longer exposures, with TRPC4 gene expression above the control at 4 h. These results suggest that altering gene expression for claudin5, TRPC4 and ZO1 by CPF may directly contribute to BBB disruption, and that the alteration is reversible upon removal of CPF.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Chlorpyrifos, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Chlorpyrifos, PESTANAL®, analytical standard