Merck
Rifaximin is an efficacious treatment for the Parkinsonian phenotype of hepatic encephalopathy.
Hepatology (Baltimore, Md.) (2013-03-09)
Beverley Kok, Matthew R Foxton, Christopher Clough, Debbie L Shawcross
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Rifaximin
Supelco
Rifaximin, VETRANAL®, analytical standard