Merck
  • Evaluation of selected biomarkers for the detection of chemical sensitization in human skin: a comparative study applying THP-1, MUTZ-3 and primary dendritic cells in culture.
Evaluation of selected biomarkers for the detection of chemical sensitization in human skin: a comparative study applying THP-1, MUTZ-3 and primary dendritic cells in culture.
Toxicology in vitro : an international journal published in association with BIBRA (2013-04-30)
Manuel Hitzler, Antje Bergert, Andreas Luch, Matthias Peiser
ABSTRAKT
Dendritic cells (DCs) exhibit the unique capacity to induce T cell differentiation and proliferation, two processes that are crucially involved in allergic reactions. By combining the exclusive potential of DCs as the only professional antigen-presenting cells of the human body with the well known handling advantages of cell lines, cell-based alternative methods aimed at detecting chemical sensitization in vitro commonly apply DC-like cells derived from myeloid cell lines. Here, we present the new biomarkers programmed death-ligand 1 (PD-L1), DC immunoreceptor (DCIR), IL-16, and neutrophil-activating protein-2 (NAP-2), all of which have been detectable in primary human DCs upon exposure to chemical contact allergens. To evaluate the applicability of DC-like cells in the prediction of a chemical's sensitization potential, the expression of cell surface PD-L1 and DCIR was analyzed. In contrast to primary DCs, only minor subpopulations of MUTZ-3 and THP-1 cells presented PD-L1 or DCIR at their surface. After exposure to increasing concentrations of nickel and cinnamic aldehyde, the expression level of PD-L1 and DCIR revealed much stronger affected on monocyte-derived DCs (MoDCs) or Langerhans cells (MoLCs) when compared to THP-1 and MUTZ-3 cells. Applying protein profiler arrays we further identified the soluble factors NAP-2, IL-16, IL-8 and MIP-1α as sensitive biomarkers showing the capacity to discriminate sensitizing from non-sensitizing chemicals or irritants. An allergen-specific release of IL-8 and MIP-1α could be detected in the supernatants of MoDCs and MoLCs and also in MUTZ-3 and THP-1 cells, though at much lower levels. On the protein and transcriptional level, NAP-2 and IL-16 indicated sensitizers most sensitively and specifically in MoDCs. Altogether, we have proven the reciprocal regulated surface molecules PD-L1 and DCIR and the soluble factors MIP-1α, NAP-2 and IL-16 as reliable biomarkers for chemical sensitization. We further show that primary DCs are significantly different in their phenotype and function compared to DC-like cell lines. Since they demonstrated higher absolute values and a broader range in biomarker expression, we propose that MoDCs represent an optimal and robust sensor test system well suited to identify and classify chemicals with an allergic potential.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Eugenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
Cinnamaldehyde, natural, ≥95%, FG
Sigma-Aldrich
trans-Cinnamaldehyde, 97%
Sigma-Aldrich
trans-Cinnamaldehyde, FCC, FG
Supelco
trans-Cinnamaldehyde, analytical standard
Sigma-Aldrich
trans-Cinnamaldehyde, ≥99%