Merck
A benzodiazepine impairs the neurogenic and behavioural effects of fluoxetine in a rodent model of chronic stress.
Neuropharmacology (2013-05-04)
Yuhui Sun, Jane Evans, Bruce Russell, Rob Kydd, Bronwen Connor
ABSTRAKT
Antidepressant agents such as fluoxetine have been shown to produce neurogenic effects involving transcriptional and translational changes that direct molecular and cellular plasticity. These cellular and molecular events appear necessary to mediate the therapeutic effects of fluoxetine and may be generated through the ability for fluoxetine to regulate BDNF levels. Clinically, benzodiazepines are frequently used in combination with standard antidepressants both for initial treatment and maintenance therapy, especially when comorbid anxiety is present. However, very little is known regarding the consequence of combined treatment of benzodiazepines and antidepressant on the development of clinical effect. The current study therefore examined the effect of co-administration of fluoxetine and the benzodiazepine, diazepam, on hippocampal neurogenesis in the social isolation rodent model of chronic stress. We demonstrate that 9 weeks of social isolation induces a deficit in motivational behaviour with increased anxiety as well as impairment in hippocampal neurogenesis. This was parallelled by reduced BDNF levels in the hippocampus. While treatment with fluoxetine alone for 3 weeks restored anxiety behaviour as well as progenitor cell proliferation and the generation of new hippocampal neurons, this effect was prevented by co-administration with diazepam. This suggests that co-administering benzodiazepines with antidepressants could significantly delay or prevent the cellular and behavioural improvement needed by patients. These findings indicate the need for future clinical studies designed to investigate the combined effects of benzodiazepines and antidepressants in patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Supelco
Diazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®