Merck
Enhancing of intensity of fluorescence by DEP manipulations of polyaniline-coated Al₂O₃ nanoparticles for immunosensing.
Biosensors & bioelectronics (2013-05-18)
Cheng-Hsin Chuang, Hsun-Pei Wu, Yao-Wei Huang, Cheng-Ho Chen
ABSTRAKT
A novel modification of low-cost Al₂O₃ nanoparticles (Al₂O₃ NPs) for antibody-protein immunosensing is proposed. The modified NPs are utilized to enhance the intensity of fluorescence in a dielectrophoretic (DEP) chip with a microelectrode array. The surface of the Al₂O₃ NPs is modified by ionic polyaniline (PANDB) rather than the conventional silane (3-aminopropyltrimethoxysilane) to conjugate the antibody on the outer shell. After the PANDB-Al₂O₃ NPs is functionalized to form probes, a DEP chip with a vertical non-uniform electric field that is produced by top and bottom electrodes condenses and immobilizes the nanoprobes on the surface of the electrodes by positive DEP force for immunosensing of the fluorescent protein. Additionally, each microelectrode array can be individually controlled with/without DEP manipulation using a computer program. Experimental results indicate that PANDB-based nanoprobes provide more rapid and sensitive immunosensing than those having undergone conventional silane modification. During immunosensing, fluorescence intensity can be doubled by the application of extra DEP force. The individual control of NPs on the microelectrode array has great potential for applications in multi-antibody arrays in a single chip for immunosensing.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, standard grade, Brockmann I, activated, basic
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, Brockmann I, standard grade, activated, acidic
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, activated, Brockmann I, standard grade, neutral
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh