Merck
Flavoring extracts of Hemidesmus indicus roots and Vanilla planifolia pods exhibit in vitro acetylcholinesterase inhibitory activities.
Plant foods for human nutrition (Dordrecht, Netherlands) (2013-05-30)
Anish Kundu, Adinpunya Mitra
ABSTRAKT
Acetylcholinesterase inhibitors (AChEIs) are important for treatment of Alzheimer's disease and other neurological disorders. Search for potent and safe AChEIs from plant sources still continues. In the present work, we explored fragrant plant extracts that are traditionally used in flavoring foods, namely, Hemidesmus indicus and Vanilla planifolia, as possible sources for AChEI. Root and pod extracts of H. indicus and V. planifolia, respectively, produce fragrant phenolic compounds, 2-hydroxy-4-methoxybenzaldehyde (MBALD) and 4-hydroxy-3-methoxybenzaldehyde (vanillin). These methoxybenzaldehydes were shown to have inhibitory potential against acetylcholinesterase (AChE). Vanillin (IC50 = 0.037 mM) was detected as more efficient inhibitor than MBALD (IC50 = 0.047 mM). This finding was supported by kinetic analysis. Thus, plant-based food flavoring agents showed capacity in curing Alzheimer's disease and other neurological dysfunctions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Supelco
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Vanillin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
2-Hydroxy-4-methoxybenzaldehyde, 98%