- A reliable method to quantify sildenafil and its metabolite N-demethylsildenafil by HPLC in plasma of children.
A reliable method to quantify sildenafil and its metabolite N-demethylsildenafil by HPLC in plasma of children.
Drug research (2013-06-06)
B Ramírez-Mendiola, C Flores-Pérez, R García-Álvarez, H Juárez-Olguín, J Flores-Pérez, J L Chávez Pacheco
PMID23737285
ABSTRAKT
The aim of this study was to develop a simple method for quantifying plasma levels of sildenafil and its metabolite by liquid chromatography with a C18 reverse-phase column and UV detection. For both compounds, linearity was assessed in the range from 10 and 1 000 ng · ml-1 and had correlation coefficients of r=0.995 and r=0.997 for sildenafil and its metabolite, respectively. The inter- and intra-day coefficients of variation was<5.3%. The limits of detection and quantification were 1 and 10 ng · ml-1. Drug levels were determined satisfactorily in two patients. A simple and reliable method was developed for use in children with Pulmonary Arterial Hypertension under treatment with sildenafil.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sildenafil solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®