Merck
Long-term bilayer encapsulation performance of atomic layer deposited Al₂O₃ and Parylene C for biomedical implantable devices.
IEEE transactions on bio-medical engineering (2013-06-12)
Xianzong Xie, Loren Rieth, Ryan Caldwell, Mohit Diwekar, Prashant Tathireddy, Rohit Sharma, Florian Solzbacher
ABSTRAKT
We present an encapsulation scheme that combines atomic layer deposited (ALD) Al₂O₃ and Parylene C for the encapsulation of implantable devices. The encapsulation performances of combining alumina and Parylene C was compared to individual layers of Parylene C or alumina and the bilayer coating had superior encapsulation properties. The alumina-Parylene coated interdigitated electrodes (IDEs) soaked in PBS for up to nine months at temperatures from 37 to 80 °C for accelerated lifetime testing. For 52-nm alumina and 6-μm Parylene C, leakage current was ∼20 pA at 5 VDC, and the impedance was about 3.5 MΩ at 1 kHz with a phase near -87° from electrochemical impedance spectroscopy for samples soaked at 67 °C for equivalent lifetime of 72 months at 37 °C. The change of impedance during the whole soaking period (up to 70 months of equivalent soaking time at 37 °C) over 1 to 10⁶ Hz was within 5%. The stability of impedance indicated almost no degradation of the encapsulation. Bias voltage effect was studied by continuously applying 5 VDC, and it reduced the lifetime of Parylene coating by ∼75% while it showed no measurable effect on the bilayer coating. Lifetime of encapsulation of IDEs with topography generated by attaching a coil and surface mount device (SMD) capacitor was about half of that of planer IDEs. The stable long-term insulation impedance, low leakage current, and better lifetime under bias voltage and topography made this double-layer encapsulation very promising for chronic implantable devices.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, standard grade, Brockmann I, activated, basic
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, Brockmann I, standard grade, activated, acidic
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, activated, Brockmann I, standard grade, neutral
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh