Merck
Mechanistic studies for the role of cellular nucleic-acid-binding protein (CNBP) in regulation of c-myc transcription.
Biochimica et biophysica acta (2013-06-19)
Siqi Chen, Lijuan Su, Jun Qiu, Nannan Xiao, Jing Lin, Jia-Heng Tan, Tian-Miao Ou, Lian-Quan Gu, Zhi-Shu Huang, Ding Li
ABSTRAKT
Guanine-rich sequence of c-myc nuclease hypersensitive element (NHE) III1 is known to fold in G-quadruplex and subsequently serves as a transcriptional silencer. Cellular nucleic-acid-binding protein (CNBP), a highly conserved zinc-finger protein with multiple biological functions, could bind to c-myc NHE III1 region, specifically to the single strand G-rich sequence. In the present study, a variety of methods, including cloning, expression and purification of protein, EMSA, CD, FRET, Ch-IP, RNA interference, luciferase reporter assay, SPR, co-immunoprecipitation, and co-transfection, were applied to investigate the mechanism for the role of CNBP in regulating c-myc transcription. We found that human CNBP specifically bound to the G-rich sequence of c-myc NHE III1 region both in vitro and in cellulo, and subsequently promoted the formation of G-quadruplex. CNBP could induce a transient decrease followed by an increase in c-myc transcription in vivo. The interaction of CNBP with NM23-H2 was responsible for the increase of c-myc transcription. Based on above experimental results, a new mechanism, involving G-quadruplex related CNBP/NM23-H2 interaction, for the regulation of c-myc transcription was proposed. These findings indicated that the regulation of c-myc transcription through NHE III1 region might be governed by mechanisms involving complex protein-protein interactions, and suggested a new possibility of CNBP as a potential anti-cancer target based on CNBP's biological function in c-myc transcription.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Nucleoside 5′-Diphosphate Kinase from bovine liver, buffered aqueous glycerol solution, ≥1,000 units/mg protein (biuret)
Sigma-Aldrich
EZ-Magna ChIP® A - Zestaw do immunoprecypitacji chromatyny, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A beads. Control primers included.