Merck
  • Temperature-dependent microwave dielectric relaxation studies of hydrogen bonded polar binary mixtures of propan-1-ol and propionaldehyde.
Temperature-dependent microwave dielectric relaxation studies of hydrogen bonded polar binary mixtures of propan-1-ol and propionaldehyde.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-06-25)
T Vishwam, K Parvateesam, S Sreeharisastry, V R K Murthy
ABSTRAKT
The molecular interaction between the polar systems of propan-1-ol and propionaldehyde for various mole fractions at different temperatures were studied by determining the frequency dependent complex dielectric permittivity by using the open-ended coaxial probe technique method in the microwave frequency range from 20 MHz to 20 GHz. The geometries are optimized at HF, B3LYP and MP2 with 6-311G and 6-311G+ basis sets. Dipole moments of the binary mixtures are calculated from the dielectric data using Higasi's method and compared with the theoretical results. Conformational analysis of the formation of hydrogen bond between the propan-1-ol and propionaldehyde is supported by the FT-IR and molecular polarizability calculations. The average relaxation times are calculated from their respective Cole-Cole plots. The activation entropy, activation enthalpy and Kirkwood correlation 'g' factor, excess permittivity (ε(E)), excess inverse relaxation time (1/τ)(E), Bruggeman parameter (f(B)) have also been determined for propan-1-ol and propionaldehyde and the results were correlated.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Propanol, suitable for HPLC, ≥99.9%
Supelco
1-Propanol, analytical standard
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
Propionaldehyde, ≥97%, FG
Sigma-Aldrich
Propionaldehyde, reagent grade, 97%