Merck
Effect of IGF-II genotype and pig rearing system on the final characteristics of dry-cured Iberian hams.
Meat science (2013-06-26)
J Sánchez Del Pulgar, A I Carrapiso, R Reina, F Biasioli, C García
ABSTRAKT
The effect of the IGF-II genotype (AG vs. GG) on the morphological and compositional parameters, the fatty acid composition of intramuscular fat, the odour concentration (analysed by dynamic olfactometry) and the volatile compound profile (analysed by proton transfer reaction time-of-flight mass spectrometry) of dry-cured Iberian ham was studied for the first time, and compared to the effect of pig rearing system (high-oleic concentrated feed vs. acorn and grass). The IGF-II genotype had no effect on most variables. However, it influenced the concentration of some odorants (methanethiol and octanal), although it did not affect odour concentration. Conversely, the rearing system had a significant effect on a large number of ham variables. Results indicate a negligible effect of the IGF-II genotype on the final ham quality and confirm that the rearing system has a marked effect.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Octanal, 99%
Sigma-Aldrich
Octanal, natural, ≥95%, FCC, FG
Sigma-Aldrich
Octanal, ≥95%, FCC, FG
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Supelco
Oleic acid, Selectophore, ≥99%
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture