Merck
The effectiveness comparison of Jitai tablets versus methadone in community-based drug treatment: a 1-year follow-up study.
Addictive behaviors (2013-07-06)
Shen-Qiang Hao, Min Zhao, Rui-Wen Zhang, Jian-Chen Zhang, Jing Zhang, Xue-Shan Feng
ABSTRAKT
The aim of the study was to compare the effectiveness of Jitai tablets (JTT) versus methadone in a community drug treatment program. A cohort study was conducted with 386 eligible subjects from 7 districts to 65 communities in Shanghai. The subjects were placed into the JTT group (n=206) or the methadone group (n=180). The data were collected at 8-, 26- and 52-week follow-ups. The retention rates of the methadone group at the 8-, 26-, and 52-week follow-ups were 97.78%, 91.67%, and 85.00%, respectively. The retention rates of the JTT group at these follow-ups were 90.78%, 83.50%, and 74.27%, respectively. A Chi-square test indicated a significant difference, and the P values were 0.0037, 0.0161, and 0.0095 for each follow-up. The relapse rates for the JTT group were 3.88%, 6.31% and 11.17% for each follow-up, and those for the methadone group were 1.11%, 2.78%, and 7.78% for each follow-up. The Chi-square test indicated no significance, and the P values were 0.1128, 0.1005 and 0.2594. A survival analysis indicated that the relapse survival curve had no significant difference between the two groups (log-rank test, P=0.188). Methadone and JTT combined with psychological intervention and social support provided effective maintenance treatment and relapse prevention in a community drug treatment program. The retention rate in the methadone group was higher, but the JTT group had the same relapse prevention as the methadone group. JTT can be recommended to clinical doctors and drug addicts.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
(±)-Methadone solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®