Merck
Paradoxical increase in survival of newborn neurons in the dentate gyrus of mice with constitutive depletion of serotonin.
The European journal of neuroscience (2013-07-12)
Silvina L Diaz, Nicolas Narboux-Nême, Sara Trowbridge, Sophie Scotto-Lomassese, Felix B Kleine Borgmann, Sebastian Jessberger, Bruno Giros, Luc Maroteaux, Evan Deneris, Patricia Gaspar
ABSTRAKT
Increased adult neurogenesis is a major neurobiological correlate of the beneficial effects of antidepressants. Indeed, selective serotonin (5-HT) re-uptake inhibitors, which increase 5-HT transmission, enhance adult neurogenesis in the dentate gyrus (DG) of the hippocampus. However, the consequences of 5-HT depletion are still unclear as studies using neurotoxins that target serotonergic neurons reached contradictory conclusions on the role of 5-HT on DG cell proliferation. Here, we analysed two genetic models of 5-HT depletion, the Pet1(-/-) and the VMAT2(f/f) ; SERT(cre/+) mice, which have, respectively, 80 and 95% reductions in hippocampal 5-HT. In both models, we found unchanged cell proliferation of the neural precursors in the DG subgranular zone, whereas a significant increase in the survival of newborn neurons was noted 1 and 4 weeks after BrdU injections. This pro-survival trait was phenocopied pharmacologically with 5-HT synthesis inhibitor PCPA treatment in adults, indicating that this effect was not developmental. Furthermore, a 1-week administration of the 5-HT1A receptor agonist 8-OH-DPAT in Pet1(-/-) and PCPA-treated mice normalised hippocampal cell survival. Overall, our results indicate that constitutive 5-HT depletion does not alter the proliferation of neural precursors in the DG but promotes the survival of newborn cells, an effect which involves activation of postsynaptic 5-HT1A receptors. The role of 5-HT in selective neuronal elimination points to a new facet in its multiple effects in controlling neural circuit maturation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Chloro-DL-phenylalanine
Sigma-Aldrich
4-Chloro-L-phenylalanine